4-Propylbenzenesulphonyl chloride 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Propylbenzene-1-sulphonyl chloride
A18354.14
A18354.14EA
147
EUR
A18354.14
4-Propylbenzenesulphonyl chloride 95%
4-Propylbenzenesulphonyl chloride
Purity: 95%
CAS No.: 146949-07-7
Molecular Formula: C9H11ClO2S
Formula Weight: 218.70
Refractive Index: 1.5375
Melting Point: 20-22°
Boiling Point: 275-276°
Beilstein Reference: 5511216
UN No.: 3265
MDL No.: MFCD00041508
Formula:
C₉H₁₁ClO₂S MW: 218,7 g/mol Boiling Pt: 275…276 °C Melting Pt: 20…22 °C Density: 1,223 Storage Temperature: Ambient |
MDL Number:
MFCD00041508 CAS Number: 146949-07-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...