6-Chloro-1-hexene ≥97%

Supplier: Thermo Fisher Scientific
Warning

H53396.06 H53396.14
H53396.06EA 78.2 EUR
H53396.06 H53396.14
6-Chloro-1-hexene ≥97%
6-Chloro-1-hexene

Purity: 97% CAS No.: 928-89-2 Molecular Formula: C6H11Cl Formula Weight: 118.61 Flash Point: 29°(84°F) Refractive Index: 1.438 Boiling Point: 135-136° UN No.: 1993 MDL No.: MFCD00039391

Formula: C₆H₁₁Cl
MW: 118.61 g/mol
Boiling Pt: 135…136 °C
Density: 0.896
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039391
CAS Number: 928-89-2
EINECS: 213-186-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR