5-Hydroxyflavone ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Hydroxy-2-phenyl-4H-chromen-4-one, 5-Hydroxy-2-phenyl-4-benzopyrone, 5-Hydroxy-2-phenylchromen-4-one, 5-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 5-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one
L14187.MC
L14187.ME
L14187.MCEA
48.2
EUR
L14187.MC
L14187.ME
5-Hydroxyflavone ≥97%
5-Hydroxyflavone
Purity: 97%
CAS No.: 491-78-1
Molecular Formula: C15H10O3
Formula Weight: 238.25
Melting Point: 156-160°
Beilstein Reference: 194429
Formula:
C₁₅H₁₀O₃ MW: 238,24 g/mol Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016944 CAS Number: 491-78-1 EINECS: 207-743-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...