Norharmane 98%

Supplier: Thermo Fisher Scientific

Synonyms: Norharman, 9H-Pyrido[3,4-b]indole

129265000 129260025 129261000
ACRO129265000EA 205 EUR
ACRO129265000 ACRO129260025 ACRO129261000
Norharmane 98%
Norharmane
Formula: C₁₁H₈N₂
Melting Pt: 199…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004956
CAS Number: 244-63-3
EINECS: 205-959-0

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 198 to 202 °C
Titration Tertiary base ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR