Butyltin hydroxide oxide 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N-butyltinhydroxideoxide, Butylhydroxyoxostannane
A18290.22
A18290.36
A18290.22EA
77.2
EUR
A18290.22
A18290.36
Butyltin hydroxide oxide 95%
Butyltin hydroxide oxide
Purity: 95%
CAS No.: 2273-43-0
Molecular Formula: CH3(CH2)3Sn(=O)OH
Formula Weight: 208.81
UN No.: 3146
MDL No.: MFCD00013927
Formula:
C₄H₁₀O₂Sn MW: 208,83 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013927 CAS Number: 2273-43-0 EINECS: 218-880-1 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...