Carbazole 96%
Supplier: Thermo Scientific
|
Warning
|
108260050
108260250
108262500
108260010
ACRO108260050EA
29
EUR
ACRO108260050
ACRO108260250
ACRO108262500
ACRO108260010
Carbazole 96%
Carbazole
Formula:
C₁₂H₉N MW: 167,21 g/mol Boiling Pt: 355 °C (760 mmHg) Melting Pt: 240…246 °C Density: 1,26 g/cm³ (20 °C) Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004960 CAS Number: 86-74-8 EINECS: 201-696-0 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Yellow to beige to light brown Crystalline powder or flakes and/or chunks |
Infrared spectrum | Conforms |
Melting point | 240 to 246 °C |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...