Carbazole, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.20255.0005 8.20255.0250
8.20255.0005EA 23.6 EUR
8.20255.0005 8.20255.0250
Carbazole, Sigma-Aldrich®
Carbazole
Formula: C₁₂H₉N
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 236 °C
Density: 1,26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004960
CAS Number: 86-74-8
EINECS: 201-696-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.1 g/l (20 °C)
Melting Point 244 - 246 °C
Molar Mass 167.2 g/mol
Bulk Density 620 kg/m3
Boiling Point 355 °C (1013 hPa)
Flash Point 220 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR