Carbazole ≥95% (by GC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
|
Warning
|
M-Clarity™ quality level = MQ200
C5132-100G
C5132-250G
C5132-500G
C5132-100GEA
91
EUR
C5132-100G
C5132-250G
C5132-500G
Carbazole ≥95% (by GC), Sigma-Aldrich®
Carbazole
Carbazole is an aromatic heterocyclic organic molecule.
- Carbazole has been used in the uronic acid assay
Carbazole mainly interacts with DNA and damages it. These events suppress the synthesis of new DNA or RNA. Carbazole derivatives are involved in antimicrobial, antitumor, antiepileptic, antihistaminic, antioxidant, anti-inflammatory, antidiarrhoeal, analgesic, neuroprotective, and pancreatic lipase suppression activities. May contain dark particles.
Formula:
C₁₂H₉N MW: 167.21 g/mol Boiling Pt: 355 °C (lit.) Melting Pt: 243...246 °C (lit.) Density: 1.1 g/cm³ (18 °C) Flash Pt: 220.0 °C (closed cup) Storage Temperature: Room Temperature |
MDL Number:
MFCD00004960 CAS Number: 86-74-8 EINECS: 201-696-0 |
|
Specification Test Results
Appearance (Colour) | White to Tan |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Carbon | 84.5 - 87.9 % |
Nitrogen | 8.2 - 8.5 % |
Purity (GC) | ≥95.0 % |
Solubility (Turbidity) 200 mg plus 4 ml of Acetone |
Clear to Slightly Hazy |
Solubility (Colour) | Colorless to Yellow |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...