Sodium cyclamate

Supplier: Roth Carl
Warning

8578.1 8578.2
ROTH8578.1EA 0 EUR
ROTH8578.1 ROTH8578.2
Sodium cyclamate
Sodium cyclamate
Formula: C₆H₁₂NNaO₃S
Melting Pt: 265 °C
MDL Number: MFCD00003827
CAS Number: 139-05-9
EINECS: 205-348-9
Merck Index: 12,02770

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR