1-Phenylethyl acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Styrallyl acetate, α-Methylbenzyl acetate
M-Clarity™ quality level = MQ200
8.43785.0100
8.43785.0500
8.43785.0100EA
124
EUR
8.43785.0100
8.43785.0500
1-Phenylethyl acetate, Sigma-Aldrich®
1-Phenylethyl acetate
Formula:
C₁₀H₁₂O₂ MW: 164,2 g/mol Boiling Pt: 94 °C (1013 hPa) Density: 1,058 g/cm³ (16 °C) |
MDL Number:
MFCD00008858 CAS Number: 93-92-5 EINECS: 202-288-5 |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | -60 °C |
Molar Mass | 164.21 g/mol |
Boiling Point | 212 °C (1013 hPa) |
Flash Point | 91 °C |
Density | 1.03 g/cm3 (20 °C) |
Ignition Point | 450 °C |
Hill Formula | C10H12O2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...