5-Chloropentyl acetate ≥98%
Supplier: Thermo Scientific
Synonyms:
Acetic acid 5-chloropentyl ester, Acetic acid 5-chloroamyl ester, 5-Chloroamyl acetate
A11111.09
A11111.18
A11111.09EA
44.4
EUR
A11111.09
A11111.18
5-Chloropentyl acetate ≥98%
5-Chloropentyl acetate
Purity: 98%
CAS No.: 20395-28-2
Molecular Formula: CH3CO2(CH2)5Cl
Formula Weight: 164.63
Flash Point: 97°(206°F)
Refractive Index: 1.4380
Boiling Point: 96-98°/11mm
Beilstein Reference: 1752107
MDL No.: MFCD00013696
Formula:
C₇H₁₃ClO₂ MW: 164,63 g/mol Boiling Pt: 96…98 °C (11 mmHg) Density: 1,061 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013696 CAS Number: 20395-28-2 EINECS: 243-784-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...