Ethyl heptafluorobutyrate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethyl 2,2,3,3,4,4,4-heptafluorobutanoate, Ethyl perfluorobutyrate
A16742.14
A16742.22
A16742.14EA
66.4
EUR
A16742.14
A16742.22
Ethyl heptafluorobutyrate ≥98%
Ethyl heptafluorobutyrate
Purity: 98%
CAS No.: 356-27-4
Molecular Formula: CF3CF2CF2CO2CH2CH3
Formula Weight: 242.09
Flash Point: 14°(57°F)
Refractive Index: 1.3020
Boiling Point: 95-98°
Beilstein Reference: 1794634
UN No.: 3272
MDL No.: MFCD00000434
Formula:
C₆H₅F₇O₂ MW: 242,09 g/mol Boiling Pt: 95…98 °C Density: 1,398 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000434 CAS Number: 356-27-4 EINECS: 206-602-1 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...