Triethylammonium bromide 99%
Supplier: Thermo Scientific
|
Warning
|
349721000
349725000
ACRO349721000EA
43.4
EUR
ACRO349721000
ACRO349725000
Triethylammonium bromide 99%
Triethylammonium bromide
Formula:
C₆H₁₅N·HBr MW: 182,1 g/mol Flash Pt: >65 °C |
MDL Number:
MFCD00054287 CAS Number: 636-70-4 EINECS: 211-263-8 |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...