2-Hexanone 98%

Supplier: Thermo Scientific
Danger

146881000 146885000 146880025
146881000.EA 227 EUR
146881000. 146885000. 146880025.
2-Hexanone 98%
2-Hexanone
Formula: C₆H₁₂O
Boiling Pt: 127 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0,81 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009482
CAS Number: 591-78-6
EINECS: 209-731-1
UN: 1224
ADR: 3,III
Merck Index: 13,06060

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3995 to 1.4015 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR