Acetonylacetone ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Hexane-2,5-dione, 2,5-Hexanedione
B25686.14
B25686.22
B25686.36
B25686.14EA
36.2
EUR
B25686.14
B25686.22
B25686.36
Acetonylacetone ≥97%
Acetonylacetone
Purity: 97%
CAS No.: 110-13-4
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 78°(172°F)
Refractive Index: 1.4250
Melting Point: -6°
Boiling Point: 192-193°
Beilstein Reference: 506525
Merck Reference: 14,71
MDL No.: MFCD00008792
Formula:
C₆H₁₀O₂ MW: 114,14 g/mol Boiling Pt: 192…193 °C Melting Pt: –6 °C Density: 0,976 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008792 CAS Number: 110-13-4 EINECS: 203-738-3 Merck Index: 14,71 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...