3,5-Heptanedione ≥98%
Supplier: Thermo Scientific
|
Warning
|
A18165.06
A18165.14
A18165.06EA
78.4
EUR
A18165.06
A18165.14
3,5-Heptanedione ≥98%
3,5-Heptanedione
Formula:
C₇H₁₂O₂ MW: 128,17 g/mol Boiling Pt: 175…177 °C Density: 0,8 Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015186 CAS Number: 7424-54-6 EINECS: 231-054-5 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...