Isoamyl acetate
Supplier: Roth Carl
|
Danger
|
Synonyms:
Isopentyl acetate, Acetic acid isoamyl ester
7341.1
7341.2
ROTH7341.1EA
85.8
EUR
ROTH7341.1
ROTH7341.2
Isoamyl acetate
Isoamyl acetate
Formula:
C₇H₁₄O₂ MW: 130,19 g/mol Boiling Pt: 142 °C (1013 hPa) Melting Pt: –78 °C Density: 0,87 g/cm³ (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008946 CAS Number: 123-92-2 EINECS: 204-662-3 UN: 1104 ADR: 3,III Merck Index: 12,05125 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...