Butyl acetate

Supplier: Roth Carl
Danger

4600.1 4600.2 4600.3 4600.4
ROTH4600.1EA 51.8 EUR
ROTH4600.1 ROTH4600.2 ROTH4600.3 ROTH4600.4
Butyl acetate
Butyl acetate
Formula: CH₃COO(CH₂)₃CH₃
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –76 °C
Density: 0,875 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR