Butyric acid

Supplier: Roth Carl
Danger

3277.1 3277.2
ROTH3277.1EA 25.2 EUR
ROTH3277.1 ROTH3277.2
Butyric acid
Butyric acid
Formula: C₄H₈O₂
Boiling Pt: 163,5 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0,958 g/cm³ (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8,III
Merck Index: 13,01593

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR