Capric acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Decylic acid, Decanoic acid
A14788.0B
A14788.0I
A14788.30
A14788.0BEA
107
EUR
A14788.0B
A14788.0I
A14788.30
Capric acid ≥99%
Capric acid
Purity: 99%
CAS No.: 334-48-5
Molecular Formula: CH3(CH2)8CO2H
Formula Weight: 172.27
Flash Point: 147°(296°F)
Physical Form: Cryst solid
Density: 0.893
Appearance: White
pH: 4
Melting Point: 29-33°
Boiling Point: 268-270°
Beilstein Reference: 1754556
Merck Reference: 14,1758
MDL No.: MFCD00004441
Formula:
C₁₀H₂₀O₂ MW: 172.27 g/mol Boiling Pt: 268…270 °C Melting Pt: 29…33 °C Density: 0.893 Flash Pt: 147 °C (296 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004441 CAS Number: 334-48-5 EINECS: 206-376-4 Merck Index: 13,01764 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...