1,2,3,4-Butanetetracarboxylic acid 99+%
Supplier: Thermo Scientific
|
Danger
|
366761000
366765000
ACRO366761000EA
75
EUR
ACRO366761000
ACRO366765000
1,2,3,4-Butanetetracarboxylic acid 99+%
1,2,3,4-Butanetetracarboxylic acid
Formula:
C₈H₁₀O₈ MW: 234,16 g/mol Boiling Pt: 460 °C (1013 hPa) Melting Pt: 195…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002722 CAS Number: 1703-58-8 EINECS: 216-938-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 188 to 192 °C |
Titration with NaOH | 99.0 to 101.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...