1,2,3,4-Butanetetracarboxylic acid ≥98%
Supplier: Thermo Scientific
|
Danger
|
A16612.22
A16612.36
A16612.22EA
66.4
EUR
A16612.22
A16612.36
1,2,3,4-Butanetetracarboxylic acid ≥98%
1,2,3,4-Butanetetracarboxylic acid
Purity: 98+%
CAS No.: 1703-58-8
Molecular Formula: C8H10O8
Formula Weight: 234.16
Melting Point: ca 190-195° dec.
Beilstein Reference: 1729167
Formula:
C₈H₁₀O₈ MW: 234,16 g/mol Boiling Pt: 460 °C (1013 hPa) Melting Pt: ∼190…195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002722 CAS Number: 1703-58-8 EINECS: 216-938-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...