Phenyl acetate 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Acetic acid phenyl ester

148770050 148771000 148770010
ACRO148770050EA 25.5 EUR
ACRO148770050 ACRO148771000 ACRO148770010
Phenyl acetate 97%
Phenyl acetate
Formula: C₈H₈O₂
Boiling Pt: 196 °C (760 mmHg)
Density: 1,07 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008699
CAS Number: 122-79-2
EINECS: 204-575-0
Merck Index: 12,07421

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4995 to 1.5035 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR