1-Chlorohexane ≥96%

Supplier: Thermo Fisher Scientific
Warning

B22379.18 B22379.30
B22379.18EA 30.7 EUR
B22379.18 B22379.30
1-Chlorohexane ≥96%
1-Chlorohexane

Purity: 96% CAS No.: 544-10-5 Molecular Formula: C6H13Cl Formula Weight: 120.62 Flash Point: 26°(78°F) Refractive Index: 1.4199 Melting Point: -94° Boiling Point: 132-134° Beilstein Reference: 1731289 Merck Reference: 14,2144 UN No.: 1993 MDL No.: MFCD00001018

Formula: C₆H₁₃Cl
MW: 120.62 g/mol
Boiling Pt: 132…134 °C
Melting Pt: –94 °C
Density: 0.878
Flash Pt: 26 °C (78 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001018
CAS Number: 544-10-5
EINECS: 208-859-5
UN: 1993
ADR: 3 III
Merck Index: 13,02163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR