1-Chlorohexane for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.18497.0005 8.18497.0250 8.18497.9022
8.18497.0005EA 22.1 EUR
8.18497.0005 8.18497.0250 8.18497.9022
1-Chlorohexane for synthesis, Sigma-Aldrich®
1-Chlorohexane
Formula: C₆H₁₃Cl
MW: 120.62 g/mol
Boiling Pt: ∼135 °C (1013 hPa)
Melting Pt: –94 °C
Density: 0.872 g/cm³ (20 °C)
Flash Pt: 27
Storage Temperature: Ambient
MDL Number: MFCD00001018
CAS Number: 544-10-5
EINECS: 208-859-5
UN: 1993
ADR: 3 II
Merck Index: 13,02163

Order Now

Specification Test Results

Solubility 0.26 g/l (20 °C)
Melting Point -94 °C
Molar Mass 120.62 g/mol
Boiling Point 132 - 136 °C (1013 hPa)
Vapor Pressure 53 hPa (55 °C)
Flash Point 27 °C
Refractive Index 1.4194 (20 °C, 589 nm)
Explosion Limit 1.0 - 9.6 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 225 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR