Perchloroethane 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hexachloroethane

148540010 148540025 148541000 148540100
ACRO148540010EA 258 EUR
ACRO148540010 ACRO148540025 ACRO148541000 ACRO148540100
Perchloroethane 99%
Perchloroethane
Formula: C₂Cl₆
MW: 236.74 g/mol
Boiling Pt: 189
Melting Pt: 184…190 °C
Density: 2.091 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000799
CAS Number: 67-72-1
EINECS: 200-666-4
UN: 3077
ADR: 9 III
Merck Index: 13,04698

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 185 to 195 °C
GC ≥98.5 %
Ash ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR