Perfluorotetradecane ≥97%

Supplier: Thermo Fisher Scientific

L16846.03 L16846.06
L16846.03EA 0 EUR
L16846.03 L16846.06
Perfluorotetradecane ≥97%
Perfluorotetradecane

Purity: 97% CAS No.: 307-62-0 Molecular Formula: C14F30 Formula Weight: 738.11 Melting Point: 103-105° MDL No.: MFCD00013571

Formula: C₁₄F₃₀
MW: 738.11 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00013571
CAS Number: 307-62-0
EINECS: 206-206-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR