1,1,1,5,5,5-Hexafluoroacetylacetone 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA
218530050
218530250
218531000
ACRO218530050EA
69.3
EUR
ACRO218530050
ACRO218530250
ACRO218531000
1,1,1,5,5,5-Hexafluoroacetylacetone 99%
1,1,1,5,5,5-Hexafluoroacetylacetone
Formula:
C₅H₂F₆O₂ MW: 208,06 g/mol Boiling Pt: 70…71 °C (760 mmHg) Density: 1,47 g/cm³ Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000426 CAS Number: 1522-22-1 EINECS: 216-191-0 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3320 to 1.3340 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...