Trifluoroacetic acid
Supplier: Roth Carl
|
Danger
|
Synonyms:
TFA
P088.1
P088.2
ROTHP088.1EA
127
EUR
ROTHP088.1
ROTHP088.2
Trifluoroacetic acid
Trifluoroacetic acid
Formula:
C₂HF₃O₂ MW: 114.02 g/mol Boiling Pt: 71.8 Melting Pt: –15 °C Density: 1.490 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8 I Merck Index: 13,09754 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...