Diiodomethane for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Methylene iodide
M-Clarity™ quality level = MQ200
8.18153.0025
8.18153.0050
8.18153.1000
8.18153.0025EA
102
EUR
8.18153.0025
8.18153.0050
8.18153.1000
Diiodomethane for synthesis, Sigma-Aldrich®
Diiodomethane
Formula:
CH₂I₂ MW: 267,84 g/mol Boiling Pt: 182 °C (1013 hPa) Melting Pt: 6 °C Density: 3,32537 g/cm³ (20 °C) Flash Pt: 110 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001079 CAS Number: 75-11-6 EINECS: 200-841-5 UN: 2810 ADR: 6.1,III Merck Index: 12,06143 |
|
Specification Test Results
Solubility | 0.8 g/l (25 °C) |
Melting Point | 6 °C |
Molar Mass | 267.84 g/mol |
Boiling Point | 181 °C (1013 hPa) (decomposition) |
Density | 3.3 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...