Trioctylamine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tri-n-octylamine

140281000 140285000 140280025
ACRO140281000EA 76 EUR
ACRO140281000 ACRO140285000 ACRO140280025
Trioctylamine 97%
Trioctylamine
Formula: C₂₄H₅₁N
MW: 353.68 g/mol
Boiling Pt: 365…367 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.809 g/cm³
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00009560
CAS Number: 1116-76-3
EINECS: 214-242-1
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4500 (20°C, 589 nm)
GC ≥96.0 %
Elemental Analysis 7.29 to 7.51 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR