Trioctylamine 95%

Supplier: Thermo Scientific
Warning

Synonyms: Tri-n-octylamine

A15067.0F A15067.AE A15067.AP
A15067.0FEA 825 EUR
A15067.0F A15067.AE A15067.AP
Trioctylamine 95%
Trioctylamine

Purity: 95% CAS No.: 1116-76-3 Molecular Formula: C24H51N Formula Weight: 353.68 Storage Temperature: 2°C to 8°C Flash Point: 158°(316°F) Refractive Index: 1.4485 Odour: Amine-like Melting Point: -39° Boiling Point: 164-168°/0.7mm Beilstein Reference: 1210618 UN No.: 3082 MDL No.: MFCD00009560

Formula: C₂₄H₅₁N
MW: 353,68 g/mol
Boiling Pt: 164…168 °C (0,7 mmHg)
Melting Pt: –39 °C
Density: 0,810
Flash Pt: 158 °C (316 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009560
CAS Number: 1116-76-3
EINECS: 214-242-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR