Butyl isocyanate for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Butyl isocyanate
M-Clarity™ quality level = MQ200
8.40099.0025
8.40099.0250
8.40099.0025EA
131
EUR
8.40099.0025
8.40099.0250
Butyl isocyanate for synthesis, Sigma-Aldrich®
Butyl isocyanate
Formula:
C₅H₉NO MW: 99,13 g/mol Boiling Pt: 112 °C (1013 hPa) Melting Pt: –75 °C Density: 0,88 g/cm³ (20 °C) Flash Pt: 19 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002046 CAS Number: 111-36-4 EINECS: 203-862-8 UN: 2485 ADR: 6.1 (3),I Merck Index: 13,01574 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...