Butylamine 99+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Butylamine, 1-Aminobutane, n-Butylamine
107800010
107800025
107800100
107802500
ACRO107800010EA
83.8
EUR
ACRO107800010
ACRO107800025
ACRO107800100
ACRO107802500
Butylamine 99+%
Butylamine
Formula:
H₃C(CH₂)₂CH₂NH₂ MW: 73,14 g/mol Boiling Pt: 78 °C (760 mmHg) Melting Pt: –49 °C Density: 0,74 g/cm³ Flash Pt: –7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011690 CAS Number: 109-73-9 EINECS: 203-699-2 UN: 1125 ADR: 3,II Merck Index: 12,01578 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4000 to 1.4020 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...