Octylamine 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Aminooctane, n-Octylamine, 1-Octylamine

B24193.22 B24193.36
B24193.22EA 28.1 EUR
B24193.22 B24193.36
Octylamine 99%
Octylamine

Purity: 99% CAS No.: 111-86-4 Molecular Formula: CH3(CH2)7NH2 Formula Weight: 129.25 Flash Point: 62°(144°F) Refractive Index: 1.4290 Melting Point: -1° Boiling Point: 178-179° Beilstein Reference: 1679227 UN No.: 2735 MDL No.: MFCD00008247

Formula: H₃C(CH₂)₆CH₂NH₂
MW: 129.25 g/mol
Boiling Pt: 178…179 °C
Melting Pt: –1 °C
Density: 0.780
Flash Pt: 60 °C (144 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008247
CAS Number: 111-86-4
EINECS: 203-916-0
UN: 2734
ADR: 8 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR