Octylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Aminooctane, n-Octylamine, 1-Octylamine
M-Clarity™ quality level = MQ200
8.06917.0250
8.06917.2500
8.06917.1000
8.06917.0250EA
53
EUR
8.06917.0250
8.06917.2500
8.06917.1000
Octylamine for synthesis, Sigma-Aldrich®
Octylamine
Formula:
H₃C(CH₂)₆CH₂NH₂ MW: 129,25 g/mol Boiling Pt: 180 °C (1013 hPa) Melting Pt: –1 °C Density: 0,7812 g/cm³ (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008247 CAS Number: 111-86-4 EINECS: 203-916-0 UN: 2734 ADR: 8 (3),I |
Specification Test Results
Solubility | 0.2 g/l (25 °C) |
Melting Point | -1 °C |
Molar Mass | 129.24 g/mol |
Boiling Point | 178 - 179 °C (1013 hPa) |
Vapor Pressure | 1 hPa (20 °C) |
Flash Point | 60 °C |
Refractive Index | 1.429 (20 °C, 589 nm) |
Explosion Limit | 1.6 - 8.2 %(V) |
Density | 0.78 g/cm3 (20 °C) |
Ignition Point | 265 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...