Pentylamine 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine

173940500 173940050 173942500
ACRO173940500EA 62.7 EUR
ACRO173940500 ACRO173940050 ACRO173942500
Pentylamine 99%
Pentylamine
Formula: H₃C(CH₂)₃CH₂NH₂
MW: 87.16 g/mol
Boiling Pt: 104.4
Melting Pt: –50 °C
Density: 0.75 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008236
CAS Number: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3 II
Merck Index: 13,00602

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
Water ≤1 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR