Pentylamine 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine

A11674.14 A11674.22 A11674.36
A11674.14EA 37 EUR
A11674.14 A11674.22 A11674.36
Pentylamine 98%
Pentylamine

Purity: 98% CAS No.: 110-58-7 Molecular Formula: CH3(CH2)4NH2 Formula Weight: 87.17 Flash Point: -1°(30°F) Refractive Index: 1.4110 Odour: Amine-like Melting Point: -55° Boiling Point: 103-105° Beilstein Reference: 505953 Merck Reference: 14,598 UN No.: 1106 MDL No.: MFCD00008236

Formula: H₃C(CH₂)₃CH₂NH₂
MW: 87.16 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –55 °C
Density: 0.751
Flash Pt: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008236
CAS Number: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3 II
Merck Index: 13,00602

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR