1-Ethylpropylamine 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3-Pentylamine, 3-Aminopentane, Pentan-3-amine
L02250.14
L02250.22
L02250.06
L02250.14EA
116
EUR
L02250.14
L02250.22
L02250.06
1-Ethylpropylamine 98%
1-Ethylpropylamine
Formula:
C₅H₁₃N MW: 87.16 g/mol Boiling Pt: 89…91 °C Density: 0.748 Flash Pt: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008096 CAS Number: 616-24-0 EINECS: 210-471-6 UN: 1106 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...