1-Docosanol 98%
Supplier: Thermo Scientific
Synonyms:
Behenyl alcohol
173720250
173721000
ACRO173720250EA
38
EUR
ACRO173720250
ACRO173721000
1-Docosanol 98%
1-Docosanol
Formula:
H₃C(CH₂)₂₀CH₂OH MW: 326,61 g/mol Boiling Pt: 376 °C (1013 hPa) Melting Pt: 68…72 °C Density: ~ 0,81 g/cm³ (20 °C) Flash Pt: 227 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002939 CAS Number: 661-19-8 EINECS: 211-546-6 Merck Index: 13,03433 |
Specification Test Results
Appearance | White Pellets or tablets |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...