Arachidyl alcohol 98%
Supplier: Thermo Scientific
Synonyms:
1-Eicosanol
215460050
215460250
ACRO215460050EA
70.3
EUR
ACRO215460050
ACRO215460250
Arachidyl alcohol 98%
Arachidyl alcohol
Formula:
H₃C(CH₂)₁₈CH₂OH MW: 298,55 g/mol Boiling Pt: 309 °C (1013 hPa) Melting Pt: 64…66 °C Density: 0,88 g/cm³ (21 °C) Flash Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002938 CAS Number: 629-96-9 EINECS: 211-119-4 |
Specification Test Results
Appearance | White Powder or pellets and/or chunks |
Infrared spectrum | Conforms |
Melting point | 62 to 66 °C |
GC | ≥97.5 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...