Methanol
|
Danger
|
Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
EMR173001
EMR173025
EURLEMR173001EA
0
EUR
EURLEMR173001
EURLEMR173025
Methanol
Methanol
Formula:
CH₃OH MW: 32,04 g/mol Boiling Pt: 64,6 °C (1013 hPa) Melting Pt: –98 °C Density: 0,7918 g/cm³ (20 °C) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...