Ethylene glycol 95%, pure

Supplier: Thermo Scientific
Warning

Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

410010010 410010025
ACRO410010010EA 39.3 EUR
ACRO410010010 ACRO410010025
Ethylene glycol 95%, pure
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62,07 g/mol
Boiling Pt: 196…198 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1,113 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=f57401712732783ab71f9c5ec1faebd0695c20bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4330 (20°C, 589 nm)
GC ≥94 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR