1-Butanethiol for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.01587.0100
8.01587.0500
8.01587.9048
8.01587.0100EA
51.4
EUR
8.01587.0100
8.01587.0500
8.01587.9048
1-Butanethiol for synthesis, Sigma-Aldrich®
1-Butanethiol
Formula:
C₄H₁₀S Boiling Pt: 98 °C (1013 hPa) Melting Pt: –116 °C Density: 0,841 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004905 CAS Number: 109-79-5 EINECS: 203-705-3 UN: 2347 ADR: 3,II Merck Index: 12,01611 |
Specification Test Results
Solubility | 0.597 g/l (20 °C) |
Melting Point | -116 °C |
Molar Mass | 90.18 g/mol |
Boiling Point | 97 - 99 °C |
Vapor Pressure | 61 hPa (25 °C) |
Flash Point | 12 °C |
Refractive Index | 1.4425 (20 °C, 589 nm) |
Explosion Limit | 1.4 - 11.3 %(V) |
Density | 0.84 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...