1-Propanethiol ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Packaged under argon.
Purity: 98%
CAS No.: 107-03-9
Molecular Formula: C3H8S
Formula Weight: 76.16
Flash Point: -20°(4°F)
Physical Form: Liquid, Packaged under argon
Density: 0.8410
Refractive Index: 1.4380
Sensitivity: Air Sensitive
Appearance: Colourless
Odour: Stench
Melting Point: -113°
Boiling Point: 67-68°
UN No.: 2402
MDL No.: MFCD00004900
Formula:
C₃H₈S Boiling Pt: 67…68 °C Melting Pt: –113 °C Density: 0,8410 Flash Pt: –20 °C (4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004900 CAS Number: 107-03-9 EINECS: 203-455-5 UN: 2402 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...