1-Propanethiol for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: n-Propanethiol, Propyl mercaptan, n-Propyl mercaptan, 1-Mercaptopropane

M-Clarity™ quality level = MQ200

8.07529.0005 8.07529.0250
8.07529.0005EA 30.5 EUR
8.07529.0005 8.07529.0250
1-Propanethiol for synthesis, Sigma-Aldrich®
1-Propanethiol
Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 67
Melting Pt: –113 °C
Density: 0.82995 g/cm³ (20 °C)
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00004900
CAS Number: 107-03-9
EINECS: 203-455-5
UN: 2402
ADR: 3 II

Order Now

Specification Test Results

Solubility 1.9 g/l (25 °C)
Melting Point -113 °C
Molar Mass 76.16 g/mol
Boiling Point 67 - 68 °C (1013 hPa)
Vapor Pressure 165 hPa (20 °C)
Flash Point -34 °C
Refractive Index 1.438 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR