Sodium octadecyl sulphate, (max. 7% H₂O) 98% (dry weight)
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Sodium Stearyl sulphate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt
A15042.03
A15042.06
A15042.14
A15042.03EA
74.8
EUR
A15042.03
A15042.06
A15042.14
Sodium octadecyl sulphate, (max. 7% H₂O) 98% (dry weight)
Sodium octadecyl sulphate
Purity: 98%
CAS No.: 1120-04-3
Molecular Formula: C18H37NaO4S
Formula Weight: 372.55
Melting Point: 176-180°
Beilstein Reference: 3640871
Formula:
C₁₈H₃₇NaO₄S Melting Pt: 176…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007469 CAS Number: 1120-04-3 EINECS: 214-295-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...