n-Docosane 99%

Supplier: Thermo Fisher Scientific

Synonyms: N-docosane

158780250 158781000 158785000
ACRO158780250EA 46.8 EUR
ACRO158780250 ACRO158781000 ACRO158785000
n-Docosane 99%
n-Docosane
Formula: H₃C(CH₂)₂₀CH₃
Boiling Pt: 369 °C (760 mmHg)
Melting Pt: 43…46 °C
Density: 0,77 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009348
CAS Number: 629-97-0
EINECS: 211-121-5

Order Now

Specification Test Results

Appearance White Crystalline solid or crystals and/or chunks
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR