n-Heneicosane 98%
Supplier: Thermo Fisher Scientific
352770100
352770500
ACRO352770100EA
158
EUR
ACRO352770100
ACRO352770500
n-Heneicosane 98%
n-Heneicosane
Formula:
H₃C(CH₂)₁₉CH₃ Boiling Pt: 100 °C (2 mmHg) Melting Pt: 39…43 °C Density: 0,7919 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009346 CAS Number: 629-94-7 EINECS: 211-118-9 |
Specification Test Results
Appearance | White Crystalline chunks or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 39 to 43 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...