n-Nonadecane 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N-nonadecane
129100250
129101000
ACRO129100250EA
41.4
EUR
ACRO129100250
ACRO129101000
n-Nonadecane 99%
n-Nonadecane
Formula:
H₃C(CH₂)₁₇CH₃ MW: 268,53 g/mol Boiling Pt: 330 °C (760 mmHg) Melting Pt: 32…34 °C Density: 0,7845 g/cm³ (20 °C) Flash Pt: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009012 CAS Number: 629-92-5 EINECS: 211-116-8 |
Specification Test Results
Appearance | White Crystalline solid |
Infrared spectrum | Conforms |
GC | ≥96 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...